SY3H3D6826B5 Synthonix Corporation

3-(Trifluoromethyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine hydrochloride

  • Cas Number : 1838669-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46559.html

Safety Information

MSDS https://synthonix.com/46559.html
Availability details for
Quantity
  • Availability
  • Add to Cart