SY3H3D6826B6 Synthonix Corporation

5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride

  • MDL number No MDL found
  • Cas Number : 115473-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/46580.html

Safety Information

MSDS https://synthonix.com/46580.html
Availability details for
Quantity
  • Availability
  • Add to Cart