SY3H3D6826B8 Synthonix Corporation

4,5,6,7-Tetrahydro-2-methyl-2H-pyrazolo[4,3-c]pyridine dihydrochloride

  • Cas Number : 157327-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46599.html

Safety Information

MSDS https://synthonix.com/46599.html
Availability details for
Quantity
  • Availability
  • Add to Cart