SY3H3D6826BA Synthonix Corporation

4-((Trifluoromethyl)thio)benzene-1,2-diamine dihydrochloride

  • Cas Number : 1313012-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46608.html

Safety Information

MSDS https://synthonix.com/46608.html
Availability details for
Quantity
  • Availability
  • Add to Cart