SY3H3D6826CD Synthonix Corporation

5,6,7-Trifluoroindoline-2,3-dione

  • MDL number No MDL found
  • Cas Number : 107583-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47226.html

Safety Information

MSDS https://synthonix.com/47226.html
Availability details for
Quantity
  • Availability
  • Add to Cart