SY3H3D6826D8 Synthonix Corporation

6-(Trifluoromethyl)benzo[d]thiazole

  • Cas Number : 131106-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47381.html

Safety Information

MSDS https://synthonix.com/47381.html
Availability details for
Quantity
  • Availability
  • Add to Cart