SY3H3D6826DA Synthonix Corporation

3-(Trifluoromethoxy)benzonitrile

  • MDL number MFCD00040959
  • Cas Number : 52771-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47453.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47453.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart