SY3H3D6826DC Synthonix Corporation

5-(Trifluoromethyl)benzo[d]oxazole

  • MDL number MFCD19374815
  • Cas Number : 1267217-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid or semi-solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47455.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47455.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart