SY3H3D6826DD Synthonix Corporation

6-(Trifluoromethoxy)benzo[d]thiazole

  • MDL number No MDL found
  • Cas Number : 876500-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47457.html

Safety Information

MSDS https://synthonix.com/47457.html
Availability details for
Quantity
  • Availability
  • Add to Cart