SY3H3D6826E1 Synthonix Corporation

1-(2,3,4,5-Tetrafluorophenyl)ethanone

  • Cas Number : 66286-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47519.html

Safety Information

MSDS https://synthonix.com/47519.html
Availability details for
Quantity
  • Availability
  • Add to Cart