SY3H3D6826EF Synthonix Corporation

1-(2,4,5-Trichlorophenyl)ethanone

  • MDL number No MDL found
  • Cas Number : 13061-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/47839.html

Safety Information

MSDS https://synthonix.com/47839.html
Availability details for
Quantity
  • Availability
  • Add to Cart