SY3H3D6826F3 Synthonix Corporation

2-Thioxo-2,3-dihydro-1H-benzo[d]imidazole-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 731742-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47895.html

Safety Information

MSDS https://synthonix.com/47895.html
Availability details for
Quantity
  • Availability
  • Add to Cart