SY3H3D6826F5 Synthonix Corporation

3-(1H-Tetrazol-5-yl)benzoic Acid

  • MDL number No MDL found
  • Cas Number : 73096-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47938.html

Safety Information

MSDS https://synthonix.com/47938.html
Availability details for
Quantity
  • Availability
  • Add to Cart