SY3H3D6826FE Synthonix Corporation

2,2,2-Trifluoro-1-(5-methylpyridin-2-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 1060801-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48111.html

Safety Information

MSDS https://synthonix.com/48111.html
Availability details for
Quantity
  • Availability
  • Add to Cart