SY3H3D682702 Synthonix Corporation

2-(Trifluoromethyl)benzohydrazide

  • Cas Number : 344-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48378.html

Safety Information

MSDS https://synthonix.com/48378.html
Availability details for
Quantity
  • Availability
  • Add to Cart