SY3H3D68270A Synthonix Corporation

1-(2,4,6-Trifluorophenyl)ethanol

  • Cas Number : 1250113-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48746.html

Safety Information

MSDS https://synthonix.com/48746.html
Availability details for
Quantity
  • Availability
  • Add to Cart