SY3H3D682720 Synthonix Corporation

1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1354940-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49489.html

Safety Information

MSDS https://synthonix.com/49489.html
Availability details for
Quantity
  • Availability
  • Add to Cart