SY3H3D682724 Synthonix Corporation

4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride

  • MDL number No MDL found
  • Cas Number : 1588441-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/49594.html

Safety Information

MSDS https://synthonix.com/49594.html
Availability details for
Quantity
  • Availability
  • Add to Cart