SY3H3D682729 Synthonix Corporation

(4-(2H-Tetrazol-5-yl)phenyl)methanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 177595-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49631.html

Safety Information

MSDS https://synthonix.com/49631.html
Availability details for
Quantity
  • Availability
  • Add to Cart