SY3H3D68272A Synthonix Corporation

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine-2-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 116118-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49649.html

Safety Information

MSDS https://synthonix.com/49649.html
Availability details for
Quantity
  • Availability
  • Add to Cart