SY3H3D68272B Synthonix Corporation

1,2,3,4-Tetrahydro-2,6-naphthyridine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 449175-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/49657.html

Safety Information

MSDS https://synthonix.com/49657.html
Availability details for
Quantity
  • Availability
  • Add to Cart