SY3H3D68272D Synthonix Corporation

(R)-1-(5-(Trifluoromethyl)pyridin-2-yl)ethanamine dihydrochloride

  • Cas Number : 2061996-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49687.html

Safety Information

MSDS https://synthonix.com/49687.html
Availability details for
Quantity
  • Availability
  • Add to Cart