SY3H3D68272F Synthonix Corporation

1-(2-Trifluoromethylphenyl)ethylamine

  • Cas Number : 273384-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49784.html

Safety Information

MSDS https://synthonix.com/49784.html
Availability details for
Quantity
  • Availability
  • Add to Cart