SY3H3D68277F Synthonix Corporation

2-(4-(Trifluoromethoxy)phenyl)ethanamine

  • Cas Number : 170015-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/49865.html

Safety Information

MSDS https://synthonix.com/49865.html
Availability details for
Quantity
  • Availability
  • Add to Cart