SY3H3D682787 Synthonix Corporation

5,6,7,8-Tetrahydroquinolin-8-ol

  • MDL number No MDL found
  • Cas Number : 14631-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50023.html

Safety Information

MSDS https://synthonix.com/50023.html
Availability details for
Quantity
  • Availability
  • Add to Cart