SY3H3D68278B Synthonix Corporation

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxamide

  • MDL number No MDL found
  • Cas Number : 57021-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50119.html

Safety Information

MSDS https://synthonix.com/50119.html
Availability details for
Quantity
  • Availability
  • Add to Cart