SY3H3D682797 Synthonix Corporation

3,4,5-Trimethylaniline

  • MDL number No MDL found
  • Cas Number : 1639-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50310.html

Safety Information

MSDS https://synthonix.com/50310.html
Availability details for
Quantity
  • Availability
  • Add to Cart