SY3H3D682799 Synthonix Corporation

5,6,7,8-Tetrahydro-1,8-naphthyridine-2-methanamine

  • MDL number No MDL found
  • Cas Number : 332883-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50324.html

Safety Information

MSDS https://synthonix.com/50324.html
Availability details for
Quantity
  • Availability
  • Add to Cart