SY3H3D68279C Synthonix Corporation

2,4,6-Trimethylbenzenesulfinamide

  • MDL number No MDL found
  • Cas Number : 137280-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50383.html

Safety Information

MSDS https://synthonix.com/50383.html
Availability details for
Quantity
  • Availability
  • Add to Cart