SY3H3D68279D Synthonix Corporation

(2,3,6-Trimethoxyphenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 380430-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50393.html

Safety Information

MSDS https://synthonix.com/50393.html
Availability details for
Quantity
  • Availability
  • Add to Cart