SY3H3D6827A0 Synthonix Corporation

3,5,5-Trimethylcyclohexane-1,2-dione

  • MDL number No MDL found
  • Cas Number : 57696-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/50436.html

Safety Information

MSDS https://synthonix.com/50436.html
Availability details for
Quantity
  • Availability
  • Add to Cart