SY3H3D6827A4 Synthonix Corporation

4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane

  • Cas Number : 1396215-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50562.html

Safety Information

MSDS https://synthonix.com/50562.html
Availability details for
Quantity
  • Availability
  • Add to Cart