SY3H3D6827AA Synthonix Corporation

4,5,6-Trichloro-2-(pyridin-3-yl)pyrimidine

  • MDL number No MDL found
  • Cas Number : 1314401-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/50645.html

Safety Information

MSDS https://synthonix.com/50645.html
Availability details for
Quantity
  • Availability
  • Add to Cart