SY3H3D6827AB Synthonix Corporation

6-Trifluoromethylisatin

  • MDL number No MDL found
  • Cas Number : 343-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50652.html

Safety Information

MSDS https://synthonix.com/50652.html
Availability details for
Quantity
  • Availability
  • Add to Cart