SY3H3D6827B0 Synthonix Corporation

5-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

  • Cas Number : 1260798-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50691.html

Safety Information

MSDS https://synthonix.com/50691.html
Availability details for
Quantity
  • Availability
  • Add to Cart