SY3H3D6827B2 Synthonix Corporation

5-(Trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 920979-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50693.html

Safety Information

MSDS https://synthonix.com/50693.html
Availability details for
Quantity
  • Availability
  • Add to Cart