SY3H3D6827B4 Synthonix Corporation

6-(6-(Trifluoromethyl)pyridin-2-yl)-1,3,5-triazine-2,4(1H,3H)-dione

  • Cas Number : 1446507-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50705.html

Safety Information

MSDS https://synthonix.com/50705.html
Availability details for
Quantity
  • Availability
  • Add to Cart