SY3H3D6827B6 Synthonix Corporation

3-(2,3,4-Trifluorophenyl)acrylic acid

  • MDL number No MDL found
  • Cas Number : 207742-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50776.html

Safety Information

MSDS https://synthonix.com/50776.html
Availability details for
Quantity
  • Availability
  • Add to Cart