SY3H3D6827B7 Synthonix Corporation

3-(2,3,6-Trifluorophenyl)acrylic acid

  • MDL number No MDL found
  • Cas Number : 230295-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50777.html

Safety Information

MSDS https://synthonix.com/50777.html
Availability details for
Quantity
  • Availability
  • Add to Cart