SY3H3D6827B8 Synthonix Corporation

2-(Trifluoromethyl)terephthalic acid

  • MDL number No MDL found
  • Cas Number : 1483-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50783.html

Safety Information

MSDS https://synthonix.com/50783.html
Availability details for
Quantity
  • Availability
  • Add to Cart