SY3H3D6827B9 Synthonix Corporation

2-(1-(Thiophen-2-yl)ethylidene)malononitrile

  • MDL number MFCD00184599
  • Cas Number : 10432-44-7
  • Molecular Weight : 174.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/50841.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/50841.html
pictograms GHS07
hstatements H317,H319
Availability details for
Quantity
  • Availability
  • Add to Cart