SY3H3D6827BC Synthonix Corporation

3-(2H-1,2,3-Triazol-4-yl)benzonitrile

  • Cas Number : 550364-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50847.html

Safety Information

MSDS https://synthonix.com/50847.html
Availability details for
Quantity
  • Availability
  • Add to Cart