SY3H3D6827C0 Synthonix Corporation

7-(Trifluoromethyl)-1H-indole

  • MDL number No MDL found
  • Cas Number : 172217-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50857.html

Safety Information

MSDS https://synthonix.com/50857.html
Availability details for
Quantity
  • Availability
  • Add to Cart