SY3H3D6827C4 Synthonix Corporation

7-(Trifluoromethoxy)-1H-indole

  • Cas Number : 396075-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50954.html

Safety Information

MSDS https://synthonix.com/50954.html
Availability details for
Quantity
  • Availability
  • Add to Cart