SY3H3D6827C8 Synthonix Corporation

(5-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl)methanol

  • MDL number No MDL found
  • Cas Number : 6758-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51133.html

Safety Information

MSDS https://synthonix.com/51133.html
Availability details for
Quantity
  • Availability
  • Add to Cart