SY3H3D6827CA Synthonix Corporation

4-[1,2,4]Triazol-1-yl-benzaldehyde

  • MDL number No MDL found
  • Cas Number : 27996-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51138.html

Safety Information

MSDS https://synthonix.com/51138.html
Availability details for
Quantity
  • Availability
  • Add to Cart