SY3H3D6827CB Synthonix Corporation

4-(2H-1,2,3-Triazol-2-yl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 179056-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/51139.html

Safety Information

MSDS https://synthonix.com/51139.html
Availability details for
Quantity
  • Availability
  • Add to Cart