SY3H3D6827CC Synthonix Corporation

5-(Thiophen-3-yl)-1H-pyrrole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 72078-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51197.html

Safety Information

MSDS https://synthonix.com/51197.html
Availability details for
Quantity
  • Availability
  • Add to Cart