SY3H3D6827D8 Synthonix Corporation

2-(4-(Trifluoromethoxy)phenoxy)acetic acid

  • Cas Number : 72220-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51309.html

Safety Information

MSDS https://synthonix.com/51309.html
Availability details for
Quantity
  • Availability
  • Add to Cart