SY3H3D6827DD Synthonix Corporation

4-(Trifluoromethyl)indoline

  • MDL number No MDL found
  • Cas Number : 905274-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51465.html

Safety Information

MSDS https://synthonix.com/51465.html
Availability details for
Quantity
  • Availability
  • Add to Cart